Assay Details
| Gene Symbol: ARL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031785.1 | NP_001026955.1 | 4 | 449 | 68 | |
| GenBank mRNA | AK392694.1 | - | 5 | 483 | 68 | |
| AK400269.1 | - | 5 | 557 | 68 | ||
| AY996811.1 | - | 4 | 449 | 68 | ||
More Information
Set Membership: |
|

ARL3
Genomic Map