Assay Details
| Gene Symbol: RAB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031787.1 | NP_001026957.1 | 6 | 549 | 72 | |
| GenBank mRNA | AK234779.1 | - | 6 | 776 | 72 | |
| AK395362.1 | - | 6 | 784 | 72 | ||
| AK400541.1 | - | 6 | 747 | 72 | ||
| AY996813.1 | - | 6 | 549 | 72 | ||
More Information
Set Membership: |
|

RAB1A
Genomic Map