Assay Details
| Gene Symbol: ADORA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032354.2 | NP_001027525.1 | 2 | 953 | 79 | |
| XM_005653722.3 | XP_005653779.1 | 1208 | 79 | |||
| XM_005653723.3 | XP_005653780.1 | 1138 | 79 | |||
| XM_005653724.3 | XP_005653781.1 | 1209 | 79 | |||
| XM_005653725.3 | XP_005653782.1 | 1072 | 79 | |||
| XM_013977657.2 | XP_013833111.2 | 1056 | 79 | |||
| XM_013977698.2 | XP_013833152.1 | 668 | 79 | |||
| XM_021102258.1 | XP_020957917.1 | 1254 | 79 | |||
| XM_021102259.1 | XP_020957918.1 | 1433 | 79 | |||
| XM_021102260.1 | XP_020957919.1 | 1316 | 79 | |||
| XM_021102261.1 | XP_020957920.1 | 1133 | 79 | |||
| XM_021102262.1 | XP_020957921.1 | 999 | 79 | |||
| XM_021102263.1 | XP_020957922.1 | 1233 | 79 | |||
| XM_021102264.1 | XP_020957923.1 | 804 | 79 | |||
| GenBank mRNA | AY772411.1 | - | 2 | 955 | 79 | |
More Information
Set Membership: |
|

ADORA1
Genomic Map