Assay Details
| Gene Symbol: ITGB1BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033009.3 | NP_001028181.1 | 11 | 943 | 117 | |
| XM_021079567.1 | XP_020935226.1 | 1138 | 117 | |||
| GenBank mRNA | AK346975.1 | - | 8 | 1496 | 117 | |
| DQ002920.2 | - | 11 | 945 | 117 | ||
| FJ447492.1 | - | 11 | 1051 | 117 | ||
More Information
Set Membership: |
|

ITGB1BP2
Genomic Map