Assay Details
| Gene Symbol: CITED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037964.1 | NP_001033053.1 | 3 | 749 | 142 | |
| XM_005657822.2 | XP_005657879.1 | 744 | 142 | |||
| XM_005657825.3 | XP_005657882.1 | 579 | 142 | |||
| XM_005657826.3 | XP_005657883.1 | 431 | 142 | |||
| XM_005657828.3 | XP_005657885.1 | 429 | 142 | |||
| XM_005657830.3 | XP_005657887.1 | 421 | 142 | |||
| XM_005657831.3 | XP_005657888.1 | 475 | 142 | |||
| XM_013990922.2 | XP_013846376.1 | 771 | 142 | |||
| XM_021079568.1 | XP_020935227.1 | 857 | 142 | |||
| XM_021079569.1 | XP_020935228.1 | 626 | 142 | |||
| XM_021079570.1 | XP_020935229.1 | 622 | 142 | |||
| XM_021079571.1 | XP_020935230.1 | 551 | 142 | |||
| XM_021079572.1 | XP_020935231.1 | 551 | 142 | |||
| XM_021079573.1 | XP_020935232.1 | 526 | 142 | |||
| XM_021079575.1 | XP_020935234.1 | 422 | 142 | |||
| GenBank mRNA | AK345373.1 | - | 4 | 591 | 141 | |
| AK348366.1 | - | 3 | 746 | 142 | ||
| DQ116787.1 | - | 3 | 749 | 142 | ||
More Information
Set Membership: |
|

CITED1
Genomic Map