Assay Details
| Gene Symbol: SSTR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038229.1 | NP_001033318.1 | 1 | 147 | 123 | |
| GenBank mRNA | AY156052.5 | - | 2 | 289 | 123 | |
| DQ234796.1 | - | 1 | 147 | 123 | ||
| DQ234798.1 | - | 1 | 147 | 123 | ||
| DQ234799.1 | - | 1 | 147 | 123 | ||
More Information
Set Membership: |
|

SSTR5
Genomic Map