Assay Details
| Gene Symbol: DNASE1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038632.1 | NP_001033721.1 | 8 | 1052 | 99 | |
| GenBank mRNA | AK237546.1 | - | 8 | 1054 | 99 | |
| AK347309.1 | - | 8 | 1036 | 99 | ||
| DQ116782.1 | - | 8 | 1052 | 99 | ||
More Information
Set Membership: |
|

DNASE1L1
Genomic Map