Assay Details
| Gene Symbol: ANGPTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038644.1 | NP_001033733.1 | 7 | 1253 | 61 | |
| GenBank mRNA | AK235723.1 | - | 7 | 1337 | 61 | |
| AK236234.1 | - | 7 | 1363 | 61 | ||
| AK398038.1 | - | 7 | 1378 | 61 | ||
| AK398906.1 | - | 7 | 1340 | 61 | ||
| AY751522.1 | - | 7 | 1253 | 61 | ||
| AY974561.1 | - | 7 | 1341 | 61 | ||
More Information
Set Membership: |
|

ANGPTL4
Genomic Map