Assay Details
| Gene Symbol: MACO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038646.1 | NP_001033735.1 | 11 | 1954 | 102 | |
| GenBank mRNA | AK393782.1 | - | 6 | 1043 | 102 | |
| AY845019.1 | - | 11 | 1954 | 102 | ||
More Information
Set Membership: |
|

MACO1
Genomic Map