Assay Details
| Gene Symbol: SUV39H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039747.1 | NP_001034836.1 | 5 | 1054 | 71 | |
| XM_005668113.3 | XP_005668170.1 | 1365 | 71 | |||
| XM_005668119.3 | XP_005668176.1 | 1343 | 71 | |||
| XM_013980236.2 | XP_013835690.1 | 1120 | 71 | |||
| GenBank mRNA | DQ400534.1 | - | 5 | 1054 | 71 | |
| EU219913.1 | - | 4 | 1026 | 71 | ||
| EU661938.1 | - | 6 | 1348 | 71 | ||
More Information
Set Membership: |
|

SUV39H2
Genomic Map