Assay Details
| Gene Symbol: PPARA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044526.1 | NP_001037991.1 | 6 | 1249 | 81 | |
| GenBank mRNA | AF228696.1 | - | 6 | 1208 | 81 | |
| DQ437887.1 | - | 6 | 1249 | 81 | ||
More Information
Set Membership: |
|

PPARA
Genomic Map