Assay Details
| Gene Symbol: FOXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044536.1 | NP_001038001.1 | 7 | 1452 | 89 | |
| XM_021098347.1 | XP_020954006.1 | 1998 | 89 | |||
| XM_021098349.1 | XP_020954008.1 | 1673 | 89 | |||
| XM_021098350.1 | XP_020954009.1 | 1702 | 89 | |||
| XM_021098351.1 | XP_020954010.1 | 1518 | 89 | |||
| XM_021098352.1 | XP_020954011.1 | 2080 | 89 | |||
| XM_021098353.1 | XP_020954012.1 | 1885 | 89 | |||
| XM_021098354.1 | XP_020954013.1 | 1653 | 89 | |||
| XM_021098355.1 | XP_020954014.1 | 1527 | 89 | |||
| XM_021098356.1 | XP_020954015.1 | 4575 | 89 | |||
| XM_021098357.1 | XP_020954016.1 | 1995 | 89 | |||
| XM_021098358.1 | XP_020954017.1 | 1640 | 89 | |||
| XM_021098359.1 | XP_020954018.1 | 1614 | 89 | |||
| XM_021098360.1 | XP_020954019.1 | 1932 | 89 | |||
| XM_021098361.1 | XP_020954020.1 | 1590 | 89 | |||
| XM_021098362.1 | XP_020954021.1 | 1668 | 89 | |||
| XM_021098363.1 | XP_020954022.1 | 1488 | 89 | |||
| XM_021098365.1 | XP_020954024.1 | 2011 | 89 | |||
| XM_021098366.1 | XP_020954025.1 | 4507 | 89 | |||
| GenBank mRNA | AB240453.1 | - | 7 | 1452 | 89 | |
| AK238909.1 | - | 6 | 1347 | 89 | ||
More Information
Set Membership: |
|

FOXN3
Genomic Map