Assay Details
| Gene Symbol: GNA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044538.2 | NP_001038003.1 | 6 | 849 | 100 | |
| GenBank mRNA | AB271926.1 | - | 5 | 682 | 100 | |
| AK392340.1 | - | 6 | 1056 | 100 | ||
| DQ256410.2 | - | 6 | 850 | 100 | ||
More Information
Set Membership: |
|

GNA11
Genomic Map