Assay Details
| Gene Symbol: CKMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044551.2 | NP_001038016.1 | 8 - 9 | 1150 | 93 | |
| XM_021082288.1 | XP_020937947.1 | 1864 | 93 | |||
| GenBank mRNA | DQ363337.1 | - | 8 - 9 | 1151 | 93 | |
More Information
Set Membership: |
|

CKMT2
Genomic Map