Assay Details
| Gene Symbol: CCR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044563.1 | NP_001038028.1 | 2 | 854 | 80 | |
| GenBank mRNA | AK391150.1 | - | 2 | 869 | 80 | |
| DQ157761.1 | - | 2 | 854 | 80 | ||
More Information
Set Membership: |
|

CCR10
Genomic Map