Assay Details
| Gene Symbol: SFN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044564.1 | NP_001038029.1 | 1 | 456 | 75 | |
| GenBank mRNA | DQ445254.1 | - | 1 | 456 | 75 | |
| KC408938.1 | - | 1 | 412 | 75 | ||
More Information
Set Membership: |
|

SFN
Genomic Map