Assay Details
| Gene Symbol: AKR1C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044569.1 | NP_001038034.1 | 7 - 8 | 849 | 102 | |
| XM_013980353.2 | XP_013835807.2 | 948 | 102 | |||
| XM_021064077.1 | XP_020919736.1 | 780 | 102 | |||
| GenBank mRNA | AK233685.1 | - | 7 - 8 | 879 | 102 | |
| AK391133.1 | - | 7 - 8 | 879 | 102 | ||
| DQ474065.1 | - | 7 - 8 | 849 | 102 | ||
| JN662505.1 | - | 7 - 8 | 879 | 102 | ||
More Information
Set Membership: |
|

AKR1C1
Genomic Map