Assay Details
| Gene Symbol: ANG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044573.2 | NP_001038038.1 | 1 | 348 | 68 | |
| XM_021098367.1 | XP_020954026.1 | 557 | 68 | |||
| GenBank mRNA | AK345883.1 | - | 2 | 767 | 68 | |
| DQ498979.1 | - | 1 | 349 | 68 | ||
| EU814501.1 | - | 2 | 460 | 68 | ||
More Information
Set Membership: |
|

ANG
Genomic Map