Assay Details
| Gene Symbol: MYOZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044593.1 | NP_001038058.1 | 6 | 846 | 81 | |
| XM_021100518.1 | XP_020956177.1 | 1052 | 81 | |||
| XM_021100520.1 | XP_020956179.1 | 983 | 81 | |||
| GenBank mRNA | DQ143038.1 | - | 6 | 846 | 81 | |
More Information
Set Membership: |
|

MYOZ2
Genomic Map