Assay Details
| Gene Symbol: MYOZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044594.1 | NP_001038059.1 | 6 | 755 | 105 | |
| XM_021082327.1 | XP_020937986.1 | 1864 | 105 | |||
| XM_021082328.1 | XP_020937987.1 | 751 | 105 | |||
| GenBank mRNA | AK396653.1 | - | 6 | 651 | 105 | |
| DQ143041.1 | - | 6 | 755 | 105 | ||
More Information
Set Membership: |
|

MYOZ3
Genomic Map