Assay Details
| Gene Symbol: MED7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044615.1 | NP_001038080.1 | 2 | 651 | 104 | |
| XM_005672584.3 | XP_005672641.1 | 905 | 104 | |||
| XM_005672585.3 | XP_005672642.1 | 719 | 104 | |||
| XM_005672586.3 | XP_005672643.1 | 970 | 104 | |||
| XM_005672587.3 | XP_005672644.1 | 868 | 104 | |||
| XM_021076588.1 | XP_020932247.1 | 641 | 104 | |||
| XM_021076589.1 | XP_020932248.1 | 994 | 104 | |||
| GenBank mRNA | AK237128.1 | - | 2 | 806 | 104 | |
| AK397215.1 | - | 2 | 917 | 104 | ||
| AY609567.1 | - | 2 | 652 | 104 | ||
| DQ200863.1 | - | 2 | 651 | 104 | ||
More Information
Set Membership: |
|

MED7
Genomic Map