Assay Details
| Gene Symbol: TRA2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077223.1 | NP_001070691.1 | 4 | 452 | 111 | |
| XM_005656690.3 | XP_005656747.1 | 656 | 111 | |||
| XM_013989583.2 | XP_013845037.1 | 1231 | 111 | |||
| XM_013989584.2 | XP_013845038.1 | 1320 | 111 | |||
| XM_013989585.2 | XP_013845039.1 | 10528 | 111 | |||
| XM_021102281.1 | XP_020957940.1 | 954 | 111 | |||
| XM_021102282.1 | XP_020957941.1 | 1044 | 111 | |||
| GenBank mRNA | AK345032.1 | - | 4 | 609 | 111 | |
| AK395816.1 | - | 4 | 616 | 111 | ||
| AY609884.1 | - | 4 | 585 | 111 | ||
| DQ972958.1 | - | 4 | 452 | 111 | ||
More Information
Set Membership: |
|

TRA2A
Genomic Map