Assay Details
| Gene Symbol: NPW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213786.1 | NP_998951.1 | 1 | 452 | 99 | |
| GenBank mRNA | AB084277.1 | - | 1 | 452 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NPW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213786.1 | NP_998951.1 | 1 | 452 | 99 | |
| GenBank mRNA | AB084277.1 | - | 1 | 452 | 99 | |
Set Membership: |
|