Assay Details
| Gene Symbol: COASY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213789.1 | NP_998954.1 | 6 | 1350 | 67 | |
| XM_013980931.2 | XP_013836385.1 | 1459 | 67 | |||
| XM_021066181.1 | XP_020921840.1 | 1463 | 67 | |||
| XM_021066182.1 | XP_020921841.1 | 1464 | 67 | |||
| GenBank mRNA | AK233026.1 | - | 6 | 1350 | 67 | |
| AK235998.1 | - | 5 | 663 | 67 | ||
| AK390700.1 | - | 5 | 691 | 67 | ||
| AY094603.1 | - | 6 | 1350 | 67 | ||
More Information
Set Membership: |
|

COASY
Genomic Map