Assay Details
| Gene Symbol: RSAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213817.1 | NP_998982.1 | 6 | 1050 | 116 | |
| GenBank mRNA | AF156929.1 | - | 6 | 1050 | 116 | |
| AK231616.1 | - | 6 | 1078 | 116 | ||
| AK347797.1 | - | 6 | 1052 | 116 | ||
| KC109004.1 | - | 6 | 1040 | 116 | ||
More Information
Set Membership: |
|

RSAD2
Genomic Map