Assay Details
| Gene Symbol: IL2RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213835.1 | NP_999000.1 | 6 | 754 | 72 | |
| XM_021064060.1 | XP_020919719.1 | 1511 | 72 | |||
| GenBank mRNA | AK346539.1 | - | 6 | 891 | 72 | |
| KF646140.1 | - | 6 | 712 | 72 | ||
| U78317.1 | - | 6 | 754 | 72 | ||
More Information
Set Membership: |
|

IL2RA
Genomic Map