Assay Details
| Gene Symbol: OXTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214027.1 | NP_999192.1 | 1 | 751 | 130 | |
| XM_005669742.3 | XP_005669799.2 | 1936 | 130 | |||
| GenBank mRNA | HM856599.1 | - | 1 | 744 | 130 | |
| X71796.1 | - | 751 | 130 | |||
More Information
Set Membership: |
|

OXTR
Genomic Map