Assay Details
| Gene Symbol: IL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214041.1 | NP_999206.1 | 3 | 348 | 87 | |
| GenBank mRNA | AK397697.1 | - | 1 | 139 | 87 | |
| EF522118.1 | - | 3 | 276 | 87 | ||
| HQ236499.1 | - | 3 | 87 | |||
| JN381193.1 | - | 3 | 87 | |||
| L20001.1 | - | 3 | 348 | 87 | ||
More Information
Set Membership: |
|

IL10
Genomic Map