Assay Details
| Gene Symbol: RAMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214089.1 | NP_999254.1 | 3 | 354 | 61 | |
| XM_021078510.1 | XP_020934169.1 | 524 | 61 | |||
| XM_021078511.1 | XP_020934170.1 | 920 | 61 | |||
| XM_021078512.1 | XP_020934171.1 | 639 | 61 | |||
| GenBank mRNA | AB090165.1 | - | 3 | 424 | 61 | |
| AY236156.1 | - | 3 | 354 | 61 | ||
More Information
Set Membership: |
|

RAMP3
Genomic Map