Assay Details
| Gene Symbol: KCNJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214151.1 | NP_999316.1 | 1 | 1153 | 79 | |
| XM_021066219.1 | XP_020921878.1 | 1548 | 79 | |||
| XM_021066220.1 | XP_020921879.1 | 1667 | 79 | |||
| XM_021066221.1 | XP_020921880.1 | 12052 | 79 | |||
| XM_021066222.1 | XP_020921881.1 | 1933 | 79 | |||
| XM_021066223.1 | XP_020921882.1 | 1845 | 79 | |||
| XM_021066224.1 | XP_020921883.1 | 11933 | 79 | |||
| GenBank mRNA | AF021140.1 | - | 1 | 1153 | 79 | |
More Information
Set Membership: |
|

KCNJ2
Genomic Map