Assay Details
| Gene Symbol: HTR1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214158.1 | NP_999323.1 | 1 | 846 | 76 | |
| XM_005653367.3 | XP_005653424.1 | 1655 | 76 | |||
| XM_013995694.2 | XP_013851148.1 | 1777 | 76 | |||
| XM_013995702.2 | XP_013851156.1 | 1774 | 76 | |||
| XM_021093548.1 | XP_020949207.1 | 1581 | 76 | |||
| GenBank mRNA | AF117655.1 | - | 1 | 846 | 76 | |
| Y11868.1 | - | 606 | 76 | |||
More Information
Set Membership: |
|

HTR1D
Genomic Map