Assay Details
| Gene Symbol: CPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214169.1 | NP_999334.1 | 11 | 1151 | 91 | |
| XM_021068540.1 | XP_020924199.1 | 1643 | 91 | |||
| GenBank mRNA | AJ133775.1 | - | 11 | 1151 | 91 | |
More Information
Set Membership: |
|

CPB1
Genomic Map