Assay Details
| Gene Symbol: DNASE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214196.1 | NP_999361.1 | 6 | 1152 | 98 | |
| GenBank mRNA | AF060221.1 | - | 6 | 1152 | 98 | |
| AJ001387.1 | - | 6 | 1086 | 98 | ||
| AK234204.1 | - | 6 | 1220 | 98 | ||
| AK390522.1 | - | 6 | 1328 | 98 | ||
| AK398356.1 | - | 5 | 919 | 98 | ||
More Information
Set Membership: |
|

DNASE2
Genomic Map