Assay Details
| Gene Symbol: PTGDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214228.1 | NP_999393.1 | 553 | 66 | ||
| GenBank mRNA | AB115763.1 | - | 226 | 66 | ||
| AK392469.1 | - | 557 | 66 | |||
| X92979.1 | - | 553 | 66 | |||
More Information
Set Membership: |
|

PTGDS
Genomic Map