Assay Details
| Gene Symbol: AVPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214232.1 | NP_999397.1 | 4 | 1054 | 124 | |
| XM_013986509.2 | XP_013841963.1 | 1193 | 124 | |||
| GenBank mRNA | X71795.1 | - | 1054 | 124 | ||
More Information
Set Membership: |
|

AVPR2
Genomic Map