Assay Details
| Gene Symbol: F12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214242.1 | NP_999407.1 | 12 | 1550 | 148 | |
| XM_021081362.1 | XP_020937021.1 | 1643 | 148 | |||
| XM_021081368.1 | XP_020937027.1 | 1661 | 148 | |||
| XM_021081376.1 | XP_020937035.1 | 119 | ||||
| XM_021081384.1 | XP_020937043.1 | 119 | ||||
| XM_021081389.1 | XP_020937048.1 | 119 | ||||
| GenBank mRNA | AB022426.1 | - | 12 | 1550 | 148 | |
| AK236881.1 | - | 4 | 612 | 148 | ||
More Information
Set Membership: |
|

F12
Genomic Map