Assay Details
| Gene Symbol: PRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214252.1 | NP_999417.1 | 1 | 151 | 80 | |
| GenBank mRNA | X16558.1 | - | 151 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PRM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214252.1 | NP_999417.1 | 1 | 151 | 80 | |
| GenBank mRNA | X16558.1 | - | 151 | 80 | ||
Set Membership: |
|