Assay Details
| Gene Symbol: PRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214253.1 | NP_999418.1 | 2 | 244 | 69 | |
| GenBank mRNA | X13381.1 | - | 244 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214253.1 | NP_999418.1 | 2 | 244 | 69 | |
| GenBank mRNA | X13381.1 | - | 244 | 69 | ||
Set Membership: |
|