Assay Details
| Gene Symbol: BCL2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214285.1 | NP_999450.1 | 1 | 552 | 72 | |
| XM_021077292.1 | XP_020932951.1 | 1620 | 72 | |||
| XM_021077293.1 | XP_020932952.1 | 772 | 72 | |||
| XM_021077294.1 | XP_020932953.1 | 869 | 72 | |||
| XM_021077295.1 | XP_020932954.1 | 1549 | 72 | |||
| XM_021077296.1 | XP_020932955.1 | 737 | 72 | |||
| XM_021077297.1 | XP_020932956.1 | 845 | 72 | |||
| XM_021077298.1 | XP_020932957.1 | 749 | 72 | |||
| GenBank mRNA | AF216205.1 | - | 1 | 521 | 72 | |
| AJ001203.1 | - | 1 | 552 | 72 | ||
More Information
Set Membership: |
|

BCL2L1
Genomic Map