Assay Details
| Gene Symbol: AMH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214310.3 | NP_999475.2 | 5 | 1529 | 64 | |
| XM_021081532.1 | XP_020937191.1 | 1603 | 64 | |||
| GenBank mRNA | AK238434.1 | - | 3 | 1277 | 64 | |
| AK238472.1 | - | 5 | 1526 | 64 | ||
| AK351175.1 | - | 5 | 1526 | 64 | ||
| AK351210.1 | - | 5 | 1530 | 64 | ||
| AK351254.1 | - | 5 | 1526 | 64 | ||
| AK351301.1 | - | 5 | 1526 | 64 | ||
| AK394912.1 | - | 4 | 1416 | 64 | ||
| U80853.1 | - | 5 | 2648 | 64 | ||
More Information
Set Membership: |
|

AMH
Genomic Map