Assay Details
| Gene Symbol: FOLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214311.1 | NP_999476.1 | 2 | 153 | 71 | |
| XM_013999190.2 | XP_013854644.1 | 1432 | 71 | |||
| GenBank mRNA | AF137374.1 | - | 2 | 153 | 71 | |
More Information
Set Membership: |
|

FOLR1
Genomic Map