Assay Details
| Gene Symbol: B3GNT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214327.1 | NP_999492.1 | 3 | 1651 | 123 | |
| XM_005670018.3 | XP_005670075.1 | 2549 | 123 | |||
| XM_013982232.2 | XP_013837686.1 | 2239 | 123 | |||
| XM_013982235.2 | XP_013837689.1 | 1691 | 123 | |||
| XM_013982236.2 | XP_013837690.1 | 1769 | 123 | |||
| XM_021068547.1 | XP_020924206.1 | 1940 | 123 | |||
| XM_021068549.1 | XP_020924208.1 | 3346 | 123 | |||
| GenBank mRNA | AY253338.1 | - | 3 | 1651 | 123 | |
More Information
Set Membership: |
|

B3GNT5
Genomic Map