Assay Details
| Gene Symbol: GATA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214328.2 | NP_999493.2 | 6 | 1704 | 62 | |
| XM_005656112.3 | XP_005656169.1 | 1936 | 62 | |||
| XM_005656113.3 | XP_005656170.1 | 2356 | 62 | |||
| XM_005656114.3 | XP_005656171.1 | 2057 | 62 | |||
| XM_021093576.1 | XP_020949235.1 | 1932 | 62 | |||
| GenBank mRNA | AF295687.1 | - | 6 | 1244 | 62 | |
More Information
Set Membership: |
|

GATA6
Genomic Map