Assay Details
| Gene Symbol: RPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214333.1 | NP_999498.1 | 10 | 1744 | 109 | |
| GenBank mRNA | AJ293582.1 | - | 10 | 1744 | 109 | |
| AK233476.1 | - | 10 | 1771 | 109 | ||
| AK239775.1 | - | 6 | 830 | 109 | ||
| AK389219.1 | - | 10 | 1734 | 109 | ||
| AK391671.1 | - | 10 | 1743 | 109 | ||
| AK400824.1 | - | 10 | 1741 | 109 | ||
More Information
Set Membership: |
|

RPN1
Genomic Map