Assay Details
| Gene Symbol: RPS15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214334.1 | NP_999499.1 | 1 | 47 | 88 | |
| GenBank mRNA | AB005624.1 | - | 1 | 47 | 88 | |
| AK231546.1 | - | 1 | 462 | 88 | ||
| AK344770.1 | - | 2 | 75 | 88 | ||
| AK347031.1 | - | 1 | 440 | 88 | ||
| AK391614.1 | - | 2 | 98 | 88 | ||
| AK391716.1 | - | 2 | 66 | 88 | ||
| AK399695.1 | - | 2 | 67 | 88 | ||
More Information
Set Membership: |
|

RPS15
Genomic Map