Assay Details
| Gene Symbol: CD180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214357.1 | NP_999522.1 | 3 | 2051 | 98 | |
| GenBank mRNA | AB190767.1 | - | 3 | 2101 | 98 | |
| AK344376.1 | - | 3 | 2070 | 98 | ||
| AY323207.1 | - | 3 | 2051 | 98 | ||
More Information
Set Membership: |
|

CD180
Genomic Map