Assay Details
| Gene Symbol: PEMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214365.1 | NP_999530.1 | 4 | 444 | 71 | |
| XM_021066249.1 | XP_020921908.1 | 627 | 71 | |||
| XM_021066250.1 | XP_020921909.1 | 632 | 71 | |||
| XM_021066251.1 | XP_020921910.1 | 630 | 71 | |||
| XM_021066252.1 | XP_020921911.1 | 473 | 71 | |||
| GenBank mRNA | AY334573.1 | - | 4 | 444 | 71 | |
More Information
Set Membership: |
|

PEMT
Genomic Map