Assay Details
| Gene Symbol: ADIPOQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214370.1 | NP_999535.1 | 3 | 749 | 77 | |
| GenBank mRNA | AK349759.1 | - | 4 | 766 | 77 | |
| AY135647.2 | - | 3 | 749 | 77 | ||
| AY589691.1 | - | 2 | 667 | 77 | ||
| EF601160.1 | - | 3 | 691 | 77 | ||
| JF730719.1 | - | 2 | 660 | 77 | ||
| JN634961.1 | - | 2 | 660 | 77 | ||
| JX092267.1 | - | 2 | 651 | 77 | ||
More Information
Set Membership: |
|

ADIPOQ
Genomic Map