Assay Details
| Gene Symbol: SUCLG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214409.2 | NP_999574.1 | 8 | 949 | 58 | |
| XM_005655225.2 | XP_005655282.2 | 999 | 58 | |||
| XM_021085885.1 | XP_020941544.1 | 1092 | 58 | |||
| GenBank mRNA | AF008588.1 | - | 8 | 899 | 58 | |
| AF008589.1 | - | 8 | 950 | 58 | ||
| AK234703.1 | - | 8 | 921 | 58 | ||
| AK239485.1 | - | 8 | 921 | 58 | ||
| AK351902.1 | - | 8 | 889 | 58 | ||
| AK392990.1 | - | 8 | 948 | 58 | ||
| AK398190.1 | - | 8 | 879 | 58 | ||
| AK398313.1 | - | 8 | 994 | 58 | ||
| AK400975.1 | - | 8 | 940 | 58 | ||
More Information
Set Membership: |
|

SUCLG1
Genomic Map