Assay Details
| Gene Symbol: CAV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348935.1 | NP_001335864.1 | 3 | 399 | 71 | |
| NM_001348936.1 | NP_001335865.1 | 3 | 374 | 71 | ||
| NM_001348937.1 | NP_001335866.1 | 3 | 360 | 71 | ||
| NM_214438.3 | NP_999603.2 | 2 | 563 | 71 | ||
| GenBank mRNA | AK234499.1 | - | 3 | 360 | 71 | |
| AK394649.1 | - | 3 | 390 | 71 | ||
| AY490204.1 | - | 3 | 352 | 71 | ||
| AY609931.1 | - | 2 | 562 | 71 | ||
| DQ835010.1 | - | 3 | 451 | 71 | ||
More Information
Set Membership: |
|

CAV1
Genomic Map